L-Methioninethylester Hydrochlorid

Lieferant: BACHEM BIOCHEMICA

BACE4003885.0100EA 0 EUR
BACE4003885.0100 BACE4003885.0025
L-Methioninethylester Hydrochlorid
L-Methioninethylester Hydrochlorid
Formel: C₇H₁₅NO₂S·HCl
Molecular Weight: 213,7 g/mol
MDL: MFCD00012508
CAS-Nummer: 2899-36-7
EINECS: 220-787-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR