Aluminiumacetat basisch
Lieferant: Honeywell Chemicals
HONE25001.9025EA
0
EUR
HONE25001.9025
Aluminiumacetat basisch
Aluminiumacetat basisch
Formel:
C₄H₇AlO₅ |
MDL:
MFCD00008688 CAS-Nummer: 142-03-0 EINECS: 205-518-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...