5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)

Lieferant: BACHEM BIOCHEMICA

BACE4031125.0250EA 99.6 EUR
BACE4031125.0250 BACEM-1795.1000
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Formel: C₈H₄BrClNNa₂O₄P
Molecular Weight: 370,43 g/mol
MDL: MFCD00036757
CAS-Nummer: 102185-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR