Magermilchpulver, Pulver, Millipore®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

SIAL70166-500GEA 43.8 EUR
SIAL70166-500G
Magermilchpulver, Pulver, Millipore®
Magermilchpulver
Lagertemperatur: Raumtemperatur
CAS-Nummer: 999999-99-4
EINECS: 310-127-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR