Milchpulver, Pulver

Lieferant: Merck

SIALBCR450-20GEA 291 EUR
SIALBCR450-20G
Milchpulver, Pulver
Milchpulver
CAS-Nummer: 999999-99-4
EINECS: 310-127-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR