Benzo[b]fluoranthen

Lieferant: LGC Standards PROMOCHEM

PROOCILULM24160.1EA 270 EUR
PROOCILULM24160.1
Benzo[b]fluoranthen
Benzo[b]fluoranthen
Formel: C₂₀H₁₂
MDL: MFCD00010582
CAS-Nummer: 205-99-2
EINECS: 205-911-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR