Benzo[j]fluoranthen

Lieferant: EHRENSTORFER GMBH

EHERC20575000EA 97 EUR
EHERC20575000
Benzo[j]fluoranthen
Benzo[j]fluoranthen
Formel: C₂₀H₁₂
CAS-Nummer: 205-82-3
EINECS: 205-910-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR