Pyridoxal-5'-phosphat Monohydrat

Lieferant: Serva

SERA33940.02EA 0 EUR
SERA33940.02 SERA33940.03
Pyridoxal-5'-phosphat Monohydrat
Pyridoxal-5'-phosphat Monohydrat
Formel: C₈H₁₀NO₆P·1H₂O
Molecular Weight: 265,17 g/mol (247,15 anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00149414
CAS-Nummer: 41468-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR