p-Xylenolblau, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Xylenol Blue
M-Clarity™ Qualitätsstufe = MQ200
205486-1G
205486-5G
205486-1GEA
30.4
EUR
205486-1G
205486-5G
p-Xylenolblau, Sigma-Aldrich®
p-Xylenolblau
Formel:
C₂₃H₂₂O₅S Molecular Weight: 410,49 g/mol |
MDL:
MFCD00005870 CAS-Nummer: 125-31-5 EINECS: 204-736-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...