1-Methyloxindol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL466921-5GEA 137 EUR
SIAL466921-5G
1-Methyloxindol, Sigma-Aldrich®
1-Methyloxindol
Formel: C₉H₉NO
Molecular Weight: 147,1739 g/mol
MDL: MFCD00030253
CAS-Nummer: 61-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR