Hydroxylammoniumchlorid 45 g/l in Ethanol, vergällt

Lieferant: Bernd Kraft

Synonyme: Hydroxylamin HydrochloridChlorhydrathydroxylaminHydroxylamin HClHydroxylamin-Hydrochlorid

KRAF20155.4700EA 0 EUR
KRAF20155.4700 KRAF20155.2700 KRAF20155.3700
Hydroxylammoniumchlorid 45 g/l in Ethanol, vergällt
Hydroxylammoniumchlorid
Formel: NH₂OH·HCl
Molecular Weight: 69,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051089
CAS-Nummer: 5470-11-1
EINECS: 226-798-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR