Dimethylglyoxim 1% in wässriger Lösung
Lieferant: Bernd Kraft
Synonyme:
Diacetylglyoxim
, Tschugaeff-Reagenz
, Diacetyldioxim
, 2,3-Butandiondioxim
, DMG
KRAF15318.3000EA
0
EUR
KRAF15318.3000
Dimethylglyoxim 1% in wässriger Lösung
Dimethylglyoxim
Formel:
CH₃C(=NOH)C(=NOH)CH₃ Molecular Weight: 116,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002117 CAS-Nummer: 95-45-4 EINECS: 202-420-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...