Dimethylglyoxim 1% in Ethanol 96% (v/v)

Lieferant: Bernd Kraft

Synonyme: DiacetylglyoximTschugaeff-ReagenzDiacetyldioxim2,3-ButandiondioximDMG

KRAF16495.3700EA 109 EUR
KRAF16495.3700
Dimethylglyoxim 1% in Ethanol 96% (v/v)
Dimethylglyoxim
Formel: CH₃C(=NOH)C(=NOH)CH₃
Molecular Weight: 116,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002117
CAS-Nummer: 95-45-4
EINECS: 202-420-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR