Tris(trimethylsilyl)phosphit

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS13860.5EA 0 EUR
SILNSILS13860.5
Tris(trimethylsilyl)phosphit
Tris(trimethylsilyl)phosphit
Formel: C₉H₂₇O₃PSi₃
Molecular Weight: 298,54 g/mol
MDL: MFCD00015588
CAS-Nummer: 1795-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR