Chloroform-[13C] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ200

SIAL485403-500MGEA 984 EUR
SIAL485403-500MG
Chloroform-[13C] (99% ¹³C ), Sigma-Aldrich®
Chloroform-[13C]
Formel: CHCl₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 31717-44-9
UN: 1888
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR