Fmoc-Met-OH

Lieferant: Molekula

MOLEM68424083EA 0 EUR
MOLEM68424083
Fmoc-Met-OH
Fmoc-Met-OH
Formel: C₂₀H₂₁NO₄S
Molecular Weight: 371,46 g/mol
MDL: MFCD00037134
CAS-Nummer: 71989-28-1
EINECS: 276-258-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR