Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)

Lieferant: Merck

47775.EA 59 EUR
47775.
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Formel: C₁₁H₈O₂
Molecular Weight: 172,19 g/mol
MDL: MFCD00001681
CAS-Nummer: 58-27-5
EINECS: 200-372-6
https://de.vwr.com/stibo/web/std.lang.all/67/17/28506717.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR