5,8,11,14-Eicosatetrainsäure

Lieferant: Cayman Chemical

CAYM90120-50EA 529 EUR
CAYM90120-50 CAYM90120-5 CAYM90120-10 CAYM90120-25
5,8,11,14-Eicosatetrainsäure
5,8,11,14-Eicosatetraynoic acid
Formel: C₂₀H₂₄O₂
Molecular Weight: 296,41 g/mol
MDL: MFCD00036967
CAS-Nummer: 1191-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR