5,7-Dichlor-8-chinolinol

Lieferant: LGC Standards PROMOCHEM

PROOMM0312.02EA 557 EUR
PROOMM0312.02
5,7-Dichlor-8-chinolinol
5,7-Dichlor-8-chinolinol
Formel: C₉H₅Cl₂NO
Molecular Weight: 214,05 g/mol
Siedepunkt: 355 °C (1013 hPa)
Schmelzpunkt: 183 °C
MDL: MFCD00006786
CAS-Nummer: 773-76-2
EINECS: 212-258-3
Merck Index: 13,02193

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR