di-Lithiumtetraborat
Lieferant: Thermo Scientific
Synonyme:
Lithiumtetraborat
96569.A1EA
0
EUR
96569.A1
96569.D9
di-Lithiumtetraborat
di-Lithiumtetraborat
Formel:
Li₂B₄O₇ Molecular Weight: 169,12 g/mol Schmelzpunkt: 918 °C Dichte: 1,4 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011083 CAS-Nummer: 12007-60-2 EINECS: 234-514-3 Merck Index: 14,05524 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...