(Bis(trifluoracetoxy)iod)benzol

Lieferant: Molekula

MOLEM64447410EA 0 EUR
MOLEM64447410
(Bis(trifluoracetoxy)iod)benzol
(Bis(trifluoracetoxy)iod)benzol
Formel: C₁₀H₅F₆IO₄
Molecular Weight: 430,04 g/mol
MDL: MFCD00009672
CAS-Nummer: 2712-78-9
EINECS: 220-308-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR