(Bis(trifluoracetoxy)iod)pentafluorbenzol

Lieferant: FLUOROCHEM LTD (CHEMPUR)

Synonyme: [Bis(trifluoroacetoxy)iodo]pentafluorobenzene

FLUO009039-1GEA 218 EUR
FLUO009039-1G
(Bis(trifluoracetoxy)iod)pentafluorbenzol
(Bis(trifluoracetoxy)iod)pentafluorbenzol
Formel: C₁₀F₁₁IO₄
Molecular Weight: 519,99 g/mol
MDL: MFCD00191612
CAS-Nummer: 14353-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR