(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 mol/l in THF, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL674656-25MLEA
306
EUR
SIAL674656-25ML
SIAL674656-5ML
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 mol/l in THF, Sigma-Aldrich®
(R)-(+)-2-Methyl-CBS-oxazaborolidine
Formel:
C₁₈H₂₀BNO Molecular Weight: 277,17 g/mol |
MDL:
MFCD00078440 CAS-Nummer: 112022-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...