N-Fmoc-L-leucin

Lieferant: Molekula

Synonyme: Fmoc-Leu-OHN-(9-Fluorenylmethoxycarbonyl)-L-leucineN-α-Fmoc-L-Leucine

MOLE15603593-25GEA 68.7 EUR
MOLE15603593-25G
N-Fmoc-L-leucin
N-Fmoc-L-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,4 g/mol
MDL: MFCD00037133
CAS-Nummer: 35661-60-0
EINECS: 252-662-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR