5-Methylthiazol
Lieferant: BEPHARM LTD.
BEPHB104923-1KGEA
0
EUR
BEPHB104923-1KG
5-Methylthiazol
5-Methylthiazole
Formel:
C₄H₅NS Molecular Weight: 99,16 g/mol Siedepunkt: 139 °C (1013 hPa) Schmelzpunkt: –40,4 °C Dichte: 1,127 g/cm³ (20 °C) |
MDL:
MFCD00005335 CAS-Nummer: 3581-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...