2-Chlor-5-cyan-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-3819-1G BB-3819-250M
COBBBB-3819-1GEA 1020 EUR
COBBBB-3819-1G COBBBB-3819-250M
2-Chlor-5-cyan-3-pyridinboronsäure
2-Chloro-5-cyano-3-pyridineboronic acid
Formel: C₆H₄BClN₂O₂
MDL: MFCD09027243
CAS-Nummer: 957060-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR