Protein & mAb Chromatography
Featured Solutions
Avantor®ACE®
Reversed-phase HPLC has become an essential tool in the separation and analysis of proteins and peptides. Industry-leading ACE® columns are available with 300 Å pores, ideal for your protein, peptide and biomoleule separations! On top of these they are available in Method Development kits consisting of 3 complementary phases for the price of a single column.