Fluorescein methotrexate triammonium salt ≥95% (durch HPLC) fluoreszierender Farbstoff

Lieferant: Biotium
Gefahr

00023
00023.EA 420 EUR
00023.
Fluorescein methotrexate triammonium salt ≥95% (durch HPLC) fluoreszierender Farbstoff
Fluorescein methotrexate triammonium salt

Fluorescein methotrexate is useful for studying anti-metabolite resistance and spontaneous gene amplification. The dye is actively taken up by cells and binds to dihydrofolate reductase (DHFR) quantitatively.


  • λEx/λEm = 496/516 nm
  • Yellow solid soluble in pH>6.5 buffer, DMF or DMSO
  • Store desiccated at 4°C and protect from light

Formel: C₄₆H₅₄N₁₄O₉S
Lagertemperatur: Kühlschrank
CAS-Nummer: 71016-04-1
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfddc0e2c0f1102474e6917f19ea4c2ea23d9734&vid=93d06cfc9e461ee3130ac1f16c463dec1e8f8d7d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR