Dysprosium ≥99,8% (REO, Basis der Oxide der Metalle der seltenen Erden), Barren
Lieferant: Thermo Fisher Scientific
00102.14
00102.22
00102.14EA
126
EUR
00102.14
00102.22
Dysprosium ≥99,8% (REO, Basis der Oxide der Metalle der seltenen Erden), Barren
Dysprosium
Purity: 99.8%
CAS No.: 7429-91-6
Physical Form: Packaged in air stable poly bottle
Odour: Odourless
MDL No.: MFCD00010982
Formel:
Dy Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010982 CAS-Nummer: 7429-91-6 EINECS: 231-073-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...