Gold ≥99,99% (Metall-Basis), Drahtform, Premion®, Ø 0.127 mm (0.005 in)

Lieferant: Thermo Fisher Scientific

Synonyme: Aurum

00130.G2 00130.G6 00130.0K
00130.G2EA 290 EUR
00130.G2 00130.G6 00130.0K
Gold ≥99,99% (Metall-Basis), Drahtform, Premion®, Ø 0.127 mm (0.005 in)
Gold

Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈0.24g/m Odour: Odourless MDL No.: MFCD00003436

Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Dichte: 19.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR