Ammoniumcarbonat, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Ammoniumcarbonat, Hirschhornsalz

0015.1000EA 124 EUR
0015.1000
Ammoniumcarbonat, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumcarbonat (enthält Ammoniumcarbamat)
Formel: (NH₄)₂CO₃
Molecular Weight: 96,09 g/mol
Schmelzpunkt: 58 °C
Dichte: 1,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010890
CAS-Nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Bestellen

Spezifikation Testergebnisse

Meets ACS Specifications
Assay (NH3) min. 30.0 %
Trace Impurities (in ppm)Chloride (Cl) max. 5
Trace Impurities (in ppm)Iron (Fe) max. 5
Trace Impurities (in ppm)Heavy Metals (as Pb) max. 5
Insoluble Matter max. 0.005 %
Non-volatile Matter max. 0.01 %
Sulfur Compounds (as SO4) max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR