di-Ammoniumhydrogencitrat, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: sek-Ammoniumcitrat, Diammoniumhydrogen-2-hydroxypropan-1,2,3-tricarboxylat, Ammoniumcitrat dibasisch, Citronensäure Diammoniumsalz

0020.1000EA 165.29 EUR
0020.1000
di-Ammoniumhydrogencitrat, BAKER ANALYZED® ACS, J.T.Baker®
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
Molecular Weight: 226.19 g/mol
Dichte: 1.483 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013068
CAS-Nummer: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Bestellen

Spezifikation Testergebnisse

Meets ACS Specifications
Assay (by formol method) 98.0 - 103.0 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.0005 %
Insoluble Matter max. 0.005 %
Oxalate (C2O4) passes test
Phosphate (PO4) max. 0.0005 %
Residue after Ignition max. 0.01 %
Sulfur Compounds (as SO4) max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR