7-Nitro-1H-indazol

Lieferant: Biotium

Synonyme: 7-NINA, 7-Nitro-1H-indazol

00240
00240.EA 211 EUR
00240.
7-Nitro-1H-indazol
7-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
Schmelzpunkt: 190 °C
Dichte: 1,56 g/cm³ (293 °C)
MDL: MFCD00022789
CAS-Nummer: 2942-42-9
EINECS: 220-934-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR