Platin ≥99,9% (Metall-Basis), Drahtform, hart, Ø 0.127 mm (0.005 in)

Lieferant: Thermo Fisher Scientific

00263.G1 00263.G5
00263.G1EA 115 EUR
00263.G1 00263.G5
Platin ≥99,9% (Metall-Basis), Drahtform, hart, Ø 0.127 mm (0.005 in)
Platinum

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.27g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179

Formel: Pt
Siedepunkt: 3827 °C (1013 hPa)
Schmelzpunkt: 1769 °C
Dichte: 21,447 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011179
CAS-Nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR