Platin ≥99,9% (Metall-Basis), Drahtform, hart, Ø 0.127 mm (0.005 in)
Lieferant: Thermo Fisher Scientific
00263.G1
00263.G5
00263.G1EA
115
EUR
00263.G1
00263.G5
Platin ≥99,9% (Metall-Basis), Drahtform, hart, Ø 0.127 mm (0.005 in)
Platinum
Purity: 99.9%
CAS No.: 7440-06-4
Physical Form: ≈0.27g/m, Temper: as drawn
Odour: Odourless
MDL No.: MFCD00011179
Formel:
Pt Siedepunkt: 3827 °C (1013 hPa) Schmelzpunkt: 1769 °C Dichte: 21,447 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011179 CAS-Nummer: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...