1,2-Diamino-9,10-anthrachinon

Lieferant: Biotium

Synonyme: 1,2-Diaminoanthraquinone, 1,2-Diamino-10-anthracenedione

00300
00300.EA 185 EUR
00300.
1,2-Diamino-9,10-anthrachinon
1,2-Diamino-9,10-anthrachinon
Formel: C₁₄H₁₀N₂O₂
Molecular Weight: 238,241 g/mol
MDL: MFCD00001219
CAS-Nummer: 1758-68-5
EINECS: 217-156-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR