Calciumacetat Hemihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Essigsäure Calciumsalz, Calciumdi(acetat)

0041.0500EA 103.35 EUR
0041.0500
Calciumacetat Hemihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Calciumacetat
Formel: (H₃CCOO)₂Ca·¹/₂H₂O
Molecular Weight: 167,18 g/mol
CAS-Nummer: 62-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR