Zirkonium ≥99,95% (Metall-Basis), Drahtform, Ø 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific

00416.BW 00416.CB
00416.BWEA 100 EUR
00416.BW 00416.CB
Zirkonium ≥99,95% (Metall-Basis), Drahtform, Ø 0.25 mm (0.01 in)
Zirkonium

Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈0.32g/m Odour: Odourless MDL No.: MFCD00011303

Formel: Zr
Lagertemperatur: Raumtemperatur
MDL: MFCD00011303
CAS-Nummer: 7440-67-7
EINECS: 231-176-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR