Bariumchlorid Dihydrat 99,0-101,0%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
0045.0100EA
33.88
EUR
0045.0100
0045.1000
Bariumchlorid Dihydrat 99,0-101,0%, BAKER ANALYZED® ACS, J.T.Baker®
Bariumchlorid Dihydrat
Formel:
BaCl₂·2H₂O Siedepunkt: 1560 °C (1013 hPa) Schmelzpunkt: 960 °C Dichte: 3,86 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149154 CAS-Nummer: 10326-27-9 EINECS: 233-788-1 UN: 1564 ADR: 6.1,III Merck Index: 14,00971 |
Spezifikation Testergebnisse
Exceeds ACS Specifications | |
Assay | 99.0 - 101.0 % |
Calcium (Ca) | max. 0.05 % |
Iron (Fe) | max. 2 ppm |
Potassium (K) | max. 0.0025 % |
Sodium (Na) | max. 0.005 % |
Strontium (Sr) | max. 0.1 % |
Heavy Metals (as Pb) | max. 5 ppm |
Insoluble Matter | max. 0.005 % |
Loss on Drying at 150°C | 14.0 - 16.0 % |
Oxidizing Substances (as NO3) | max. 0.005 % |
pH of 5% Solution at 25°C | 5.2 - 8.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...