Quecksilber 99,999 Metals basis ACS

Lieferant: Thermo Fisher Scientific
Gefahr

00522.XA 00522.TD 00522.TC 00522.XB
00522.XAEA 505 EUR
00522.XA 00522.TD 00522.TC 00522.XB
Quecksilber 99,999 Metals basis ACS
Mercury

Purity: 99.999% CAS No.: 7439-97-6 Physical Form: Triple distilled Appearance: Silver Odour: Odourless UN No.: 2809 MDL No.: MFCD00011035

Formel: Hg
Molecular Weight: 200.59 g/mol
Siedepunkt: 356.5
Schmelzpunkt: –39 °C
Dichte: 13.54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011035
CAS-Nummer: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8 III
Merck Index: 13,05925

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR