di-Ammoniumhydrogenphosphat, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Synonyme:
di-Ammoniumorthophosphat, Ammoniumphosphat dibasisch, di-Ammoniumphosphat, Diammoniumhydrogenphosphat, sek. Ammoniumphosphat, Ammoniumorthophosphat dibasisch
0054.1000EA
108.31
EUR
0054.1000
di-Ammoniumhydrogenphosphat, BAKER ANALYZED® ACS, J.T.Baker®
di-Ammoniumhydrogenphosphat
Formel:
(NH₄)₂HPO₄ Molecular Weight: 132,06 g/mol |
MDL:
MFCD00010891 CAS-Nummer: 7783-28-0 |
Spezifikation Testergebnisse
Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs. | |
Assay | min. 98.0 % |
Calcium (Ca) | max. 0.001 % |
Iron (Fe) | max. 0.001 % |
Potassium (K) | max. 0.005 % |
Magnesium (Mg) | max. 0.0005 % |
Sodium (Na) | max. 0.005 % |
Heavy Metals (as Pb) | max. 0.001 % |
Insoluble Matter | max. 0.005 % |
Nitrate (NO3) | max. 0.003 % |
pH of 5% Solution at 25°C | 7.7 - 8.1 |
Sulfate (SO4) | max. 0.01 % |
Trace Impurities (in ppm)Chloride (Cl) | max. 5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...