Praseodym ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.62 mm (0.024 in)

Lieferant: Thermo Fisher Scientific

00607.FF
00607.FFEA 386 EUR
00607.FF
Praseodym ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 0.62 mm (0.024 in)
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Physical Form: ≈2.6g/25x25mm, Packaged in mylar under argon Appearance: Grey Odour: Odourless MDL No.: MFCD00011174

Formel: Pr
Molecular Weight: 140.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR