Silber ≥99,95% (Metall-Basis), Pulver -100 mesh
Lieferant: Thermo Fisher Scientific
00690.14
00690.22
00690.14EA
203
EUR
00690.14
00690.22
Silber ≥99,95% (Metall-Basis), Pulver -100 mesh
Silver
Purity: 99.95%
CAS No.: 7440-22-4
Appearance: Silver
Odour: Characteristic
UN No.: 3077
MDL No.: MFCD00003397
Formel:
Ag Siedepunkt: 2212 °C (1013 hPa) Schmelzpunkt: 961,9 °C Dichte: 10,5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003397 CAS-Nummer: 7440-22-4 EINECS: 231-131-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...