ortho-Borsäure 99,5-100,5% (Trockenmasse), körnig NF, J.T.Baker®

Lieferant: Avantor
Gefahr

0091-05EA 312.41 EUR
0091-05 0091-01
ortho-Borsäure 99,5-100,5% (Trockenmasse), körnig NF, J.T.Baker®
ortho-Borsäure
Formel: B(OH)₃
Siedepunkt: 185 °C (1013 hPa)
Schmelzpunkt: 160 °C
Dichte: 1,44 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011337
CAS-Nummer: 10043-35-3
EINECS: 233-139-2
ADR: 5.1,III
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Meets N.F. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤0.5%
Bulk Pharmaceutical Chemical 99.5 - 100.5%
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Solubility in Alcohol Passes Test
NF - Loss on Drying ≤0.5 %
NF - Assay (H₃BO₃) (dried basis) 99.5 - 100.5 %
Not for Internal Use
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR