HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem® zur Synthese

Lieferant: NOVABIOCHEM
Gefahr

Synonyme: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)

M-Clarity™ Qualitätsstufe = MQ300

8.51006.0100 8.51006.0025 8.51006.1000
01-62-0010-100EA 182 EUR
01-62-0010-100 01-62-0010-25 8.51006.1000
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem® zur Synthese
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel: C₁₁H₁₆F₆N₅OP
Molecular Weight: 379.2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00075445
CAS-Nummer: 94790-37-1
EINECS: 423-020-5
UN: 1325
ADR: 4.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR