tri-Natriumcitrat Dihydrat ≥99%, Reagenzqualität

Lieferant: VWR Chemicals

0101-1KGEA 84.9 EUR
0101-1KG 0101-2.5KG 0101-500G 0101-12KG 0101-50KG
tri-Natriumcitrat Dihydrat ≥99%, Reagenzqualität
tri-Natriumcitrat Dihydrat

A biological buffer used for cell culture, in vitro, enzyme assays and some electrophoretic applications at physiological pH.

Formel: Na₃C₆H₅O₇·2H₂O
Schmelzpunkt: 150 °C
Dichte: 1,76 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Calcium ≤ 0,005%
Chloride ≤ 0,005%
DNase none detected
Heavy Metals (as Pb) ≤0,001%
Insolubles ≤0,005%
Iron ≤0,0005%
Loss on Drying (%) 10,0 - 13,0
pH (5%, Wasser)@25°C 7,0 - 9,0
Protease none detected
Purity (Dry Basis) (%) ≥ 99%
RNase none detected
Sulphate ≤0,005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR