Citronensäure Monohydrat 99,5-100,5% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

0116-07EA 996.8 EUR
0116-07
Citronensäure Monohydrat 99,5-100,5% (durch Trockensubstanz), körnig USP, Multikompendial, J.T.Baker®
Citronensäure Monohydrat
Formel: C₆H₈O₇.H₂O
Molecular Weight: 210,14 g/mol
Schmelzpunkt: 135…152 °C
Dichte: 1,54 g/cm³
Flammpunkt: 173,9 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149972
CAS-Nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.5 - 100.5%
Meets B.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications Passes Test
Meets J.P. Chemical Specifications Passes Test
Meets U.S.P Requirements ≤0.036%
Preserve in Tight Containers Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤0.1%
Bulk Pharmaceutical Chemical ≤0.015%
USP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification Passes Test
USP - Oxalic Acid ≤ 0.036 %
USP - Readily Carbonizable Substances Passes Test
USP - Residue on Ignition ≤ 0.1 %
USP - Sulfate (SO₄) ≤ 0.015 %
USP - Water (H₂O) 7.5 - 9.0 %
EP/BP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Ash (sulfated) ≤ 0.1 %
EP/BP - Endotoxin Concentration, IU/mg ≤ 0.5
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Oxalic Acid ≤ 360 ppm
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Sulfate (SO₄) ≤ 150 ppm
EP/BP - Water (H₂O) 7.5 - 9.0 %
JP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis) 99.5 - 100.5 %
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Identification Passes Test
JP - Oxalic Acid ≤ 360 ppm
JP - Water (H₂O) 7.5 - 9.0 %
JP - Readily Carbonizable Substances Passes Test
JP - Residue on Ignition ≤ 0.10 %
JP - Sulfate (SO₄) ≤ 150 ppm
ACS - Iron (Fe) ≤ 3 ppm
Lead (Pb) ≤ 2 ppm
Must be subjected to further processing during the preparation of
injectable dosage forms.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP <232>, EP 5.20) - Information on
elemental impurities for this product is available on the associated
Product Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR