Citronensäure, wasserfrei 99,5-100,5% (durch Trockensubstanz), Pulver USP, J.T.Baker®

Lieferant: Avantor
Achtung

0122-01EA 83.99 EUR
0122-01 0122-05 0122-07 0122-R
Citronensäure, wasserfrei 99,5-100,5% (durch Trockensubstanz), Pulver USP, J.T.Baker®
Citronensäure

Standards: All the products are cGMP produced and meet USP grade standards.

Formel: HOC(COOH)(CH₂COOH)₂
Siedepunkt: 310 °C (1013 hPa)
Schmelzpunkt: 149…151 °C
Dichte: 1,44 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011669
CAS-Nummer: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.5 - 100.5%
Meets U.S.P Requirements Passes Test
Preserve in Tight Containers Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.1%
USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Oxalic Acid ≤0.036 %
USP - Sulfate (SO₄) ≤0.015 %
USP - Readily Carbonizable Substances Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
Must be subjected to further processing during the preparation of
injectable dosage forms
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Meets Reagent Specifications for testing USP/NF monographs
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR