Eisen(III)sulfat Hydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Achtung

0123.1000EA 124 EUR
0123.1000
Eisen(III)sulfat Hydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Eisen(III)sulfat Hydrat
Formel: Fe₂(SO₄)₃·xH₂O
Dichte: 3,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149714
CAS-Nummer: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Bestellen

Spezifikation Testergebnisse

Assay (Fe2(SO4)3) min. 73.0 %
Chloride (Cl) max. 0.002 %
Copper (Cu) max. 0.005 %
Zinc (Zn) max. 0.005 %
Ferrous Iron (as Fe) max. 0.02 %
Insoluble Matter max. 0.02 %
Nitrate (NO3) max. 0.01 %
Substances not Precipitated by NH4OH (as SO4) max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR