Ameisensäure 88%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
0128-01EA
156
EUR
0128-01
Ameisensäure 88%, BAKER ANALYZED® ACS, J.T.Baker®
Ameisensäure
Formel:
HCO₂H Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003297 CAS-Nummer: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8, 3,II Merck Index: 13,04265 |
|
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | ≥88.0% |
For Laboratory,Research,or Manufacturing Use | ≤15 |
Assay (HCOOH) | ≥88.0 % |
Color (APHA) | ≤15 |
Dilution Test | Passes Test |
Residue after Evaporation | ≤0.0020 % |
Acetic Acid (CH₃COOH) | ≤0.4 % |
Ammonium (NH₄) | ≤0.005 % |
Sulfate (SO₄) | ≤0.002 % |
Sulfite (SO₃) | Passes Test |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...